Learn More
CD107a Rat anti-Mouse, FITC, Clone: 1D4B, BD
Descripción
The 1D4B antibody recognizes CD107a (Lysosome-Associated Membrane Protein 1, LAMP-1), one of the two major glycoproteins in lysosome membranes which are useful markers to distinguish lysosomes from other organelles. Mouse CD107a consists of a 40kDa core protein which is heavily glycosylated to form heterogeneous mature glycoprotein of 110-140kDa. It is principally expressed in epithelial cells and macrophages in a variety of organs in normal and Beige mutant mice. Several lines of evidence suggest that cell-surface mouse and human LAMP-1 glycoproteins, the expression of which may correlate with malignant transformation, participate in intercellular adhesion and adhesion to extracellular matrix.
Host Species: Rat
Clone: 1D4B
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Plasma membrane fraction of mouse embryo NIH 3T3 cell line
Intracellular Staining
Especificaciones
Especificaciones
| Antígeno | CD107a |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 1D4B |
| Concentración | 0.5mg/mL |
| Conjugado | FITC |
| Descripción | LAMP-1; LGP-120; LGP-A; P2B |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Especie del huésped | Rat |
| Inmunógeno | Plasma membrane fraction of mouse embryo NIH 3T3 cell line |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.