Learn More
CD104 Mouse anti-Human, Unlabeled, Clone: 7, BD
Descripción
Cell adhesion to extracellular matrix components or to cell surface proteins, especially those expressed by leukocytes and endothelial cells, is mediated by integrins. Integrins contain noncovalently associated α and β subunits. At least 17 α and 8 β subunits have been identified and these proteins can heterodimerize to form 22 different receptors. The α6β4 integrin is a receptor for various laminins and binds with the highest affinity to laminins 4 and 5. It exhibits elevated expression in the basal cell layer of stratified epithelia, in Schwann cells at the onset of myelination, and in CD4-CD8- pre-T lymphocytes entering the thymus. In addition, α6β4 expression is increased in squamous carcinomas where it promotes invasion through a targeting of PI3 kinase activity. The majority of β4 comprises a cytoplasmic domain with unique signaling properties. The C-terminal portion of this domain contains two pairs of type III fibronectin-like motifs (FNIII) and a tyrosine activation motif (TAM). Additional domains in the cytoplasmic tail bind Shc and activate the MAPK pathway. Thus, integrin β4 is an integrin subunit that is important for cell survival, growth, and differentiation.
Host Species: Mouse
Clone: 7
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Integrin β4 aa. 1612-1821
Formula Weight [Chemical]: 200kDa
Immunofluorescence, Western Blotting
Especificaciones
Especificaciones
| Antígeno | CD104 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 7 |
| Concentración | 250μg/mL |
| Conjugado | Unconjugated |
| Descripción | Integrin β4 |
| Formulación | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human Integrin β4 aa. 1612-1821 |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.