missing translation for 'onlineSavingsMsg'
Learn More

δ-Catenin Mouse, Unlabeled, Clone: 30, BD

Código de producto. 15815969
Change view
Click to view available options
Cantidad:
150 μg
50 μg
Tamaño de la unidad:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15815969 50 μg 1 set
15825969 150 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15815969 Proveedor BD Biosciences N.º de proveedor 611536

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The catenins (α, β, γ, δ, p120[ctn]) are cytoplasmic proteins that are related to the Drosophila Armadillo protein. Catenins may have dual roles since they are components of cell-cell adherens junctions and can translocate to the nucleus after stimulation of the Wingless (Wnt-1 homolog) signaling pathway. α-Catenin has two subtypes: αE-Catenin, which is expressed ubiquitously, and αN-Catenin, which is expressed in the nervous system. β-Catenin binds to the cytoplasmic tail of E-Cadherin at adherens junctions and has been implicated in Wnt-1 signaling. γ-Catenin is associated with desmoglein in desmosomes and is closely related to β-Catenin. p120[ctn] is related to both β- and γ-catenin and is a substrate of tyrosine kinases localized at adherens junctions. δ-catenin was identified by its ability to bind the Alzheimer's disease-related protein, presenilin-1. It is most closely related to p120[ctn] and the desmosomal protein, p0071. It contains 10 Armadillo (Arm) repeats, as compared to the 13 Arm repeats found in β-Catenin. δ-Catenin is expressed at high levels in the developing nervous system, where it may be involved in neuronal progenitor cell migration and dendrite development.

Host Species: Mouse
Clone: 30
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse δ-Catenin aa. 85-194
Formula Weight [Chemical]: 160kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno δ-Catenin
Aplicaciones Western Blot
Clasificación Monoclonal
Clon 30
Concentración 250μg/mL
Conjugado Unconjugated
Formulación Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Mouse δ-Catenin aa. 85-194
Método de purificación Affinity Purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Mouse, Rat
Contenido y almacenamiento Store undiluted at -20°C.
Isotype IgG1
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.