Learn More
Caspase-8 Mouse anti-Human, Unlabeled, Clone: 4-1-20, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 551244
Descripción
Caspase-8 (FLICE/MACH-1) is a 55 kDa cytosolic protein with homology to the CD95/Fas-associated signal transducer, FADD/MORT-1, as well as to other caspase (ICE/Ced-3) cysteine proteases. The N-terminal region of caspase-8 contains an amino acid sequence, termed the death domain, that facilitates caspase-8-FADD direct interaction. FADD therefore acts as an adapter molecule, allowing caspase-8 to become recruited to the cytoplasmic region of Fas following receptor activation. Viral proteins (v-FLIPS) which inhibit recruitment and activation of caspase-8 have been isolated. Caspase-8 is produced as a proenzyme (55/50 kDa doublet) which upon receptor aggregation is proteolytically cleaved into smaller subunits of 40/36 (doublet), and 23 kDa. Overexpression of caspase-8 is sufficient to induce apoptosis in certain cell lines (e.g., MCF-7) and this phenotype is blocked by overexpression of the caspase-3 protease inhibitor, CrmA. The antibody recognizes both the proform of human caspase-8 (55/50 kDa doublet) as well as the cleaved form (40/36 kDa doublet) on SDS/PAGE. Full-length recombinant human caspase-8 protein was used as immunogen.
Especificaciones
| Caspase-8 | |
| Monoclonal | |
| 0.5mg/mL | |
| FLICE, MACH-1, Mch5 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Western Blot | |
| 4/1/2020 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human caspase-8 recombinant protein | |
| 50 μg | |
| Cell Biology | |
| Human | |
| IgG1 κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.