Learn More
CAPN3, Mouse, Clone: 1B2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CAPN3.
Marca: Abnova H00000825-M01.100ug
Descripción
Calpain, a heterodimer consisting of a large and a small subunit, is a major intracellular protease, although its function has not been well established. This gene encodes a muscle-specific member of the calpain large subunit family that specifically binds to titin. Mutations in this gene are associated with limb-girdle muscular dystrophies type 2A. Alternate promoters and alternative splicing result in multiple transcript variants encoding different isoforms and some variants are ubiquitously expressed. [provided by RefSeq
Sequence: NKIKAWQKIFKHYDTDQSGTINSYEMRNAVNDAGFHLNNQLYDIITMRYADKHMNIDFDSFICCFVRLEGMFRAFHAFDKDGDGIIKLNVLEWLQLTMYAEspecificaciones
| CAPN3 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC003169 | |
| CAPN3 | |
| CAPN3 (AAH03169.1, 210 a.a. ∼ 309 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunofluorescence, Western Blot | |
| 1B2 | |
| Mouse monoclonal antibody raised against a partial recombinant CAPN3. | |
| CAPN3 | |
| CANP3/CANPL3/LGMD2/LGMD2A/MGC10767/MGC11121/MGC14344/MGC4403/nCL-1/p94 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 825 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.