Learn More
CaMKK beta Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA530558
Descripción
PA5-30558 targets CaMKK beta in IF and WB applications and shows reactivity with Human and mouse samples. The PA5-30558 immunogen is recombit fragment corresponding to a region within amino acids 231 and 525 of Human CaMKK beta. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
The product of this gene belongs to the Serine/Threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. This protein plays a role in the calcium/calmodulin-dependent (CaM) kinase cascade by phosphorylating the downstream kinases CaMK1 and CaMK4. Seven transcript variants encoding six distinct isoforms have been identified for this gene. Additional splice variants have been described but their full-length nature has not been determined. The identified isoforms exhibit a distinct ability to undergo autophosphorylation and to phosphorylate the downstream kinases.
Especificaciones
| CaMKK beta | |
| Polyclonal | |
| Unconjugated | |
| Q96RR4, Q8C078 | |
| CAMKK2 | |
| Recombit fragment corresponding to a region within amino acids 231 and 525 of Human CaMKK beta | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| CAMKK2 | |
| CAMKK, CAMKKB, CAMKK beta protein, caM-KK 2, caM-KK beta, caM-kinase kinase 2, caM-kinase kinase beta, calcium/calmodulin-dependent protein kinase beta, calcium/calmodulin-dependent protein kinase kinase 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10645, 207565 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.