missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CA1, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marca: Abnova H00000759-PW1.1Set
Includes: Antibody pair for IP: mouse monoclonal anti-CA1 (300μg), Antibody pair for WB: rabbit polyclonal anti-CA1 (50μL)
Especificaciones
| CA1 | |
| Monoclonal/Polyclonal | |
| This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
| Car1 | |
| Mouse, Rabbit | |
| RUO | |
| 759 | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| CA1 | |
| CA1 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-CA1 (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-CA1 (50 ul) | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido