Learn More
C3, Mouse, Clone: 5F9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant C3.
Marca: Abnova H00000718-M01A.200uL
Descripción
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection. [provided by RefSeq
Sequence: DKACEPGVDYVYKTRLVKVQLSNDFDEYIMAIEQTIKSGSDEVQVGQQRTFISPIKCREALKLEEKKHYLMWGLSSDFWGEKPNLSYIIGKDTWVEHWPEEDECQDEENQKEspecificaciones
| C3 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_000064 | |
| C3 | |
| C3 (NP_000055, 1534 a.a. ∼ 1644 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 718 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 5F9 | |
| Mouse monoclonal antibody raised against a partial recombinant C3. | |
| C3 | |
| ARMD9/ASP/CPAMD1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.