Learn More
baculoviral IAP repeat-containing 5, Mouse, Clone: 5B10, Abnova™
Descripción
This gene is a member of the inhibitor of apoptosis (IAP) gene family, which encode negative regulatory proteins that prevent apoptotic cell death. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but this gene encodes proteins with only a single BIR domain. The encoded proteins also lack a C-terminus RING finger domain. Gene expression is high during fetal development and in most tumors yet low in adult tissues. Antisense transcripts are involved in the regulation of this gene's expression. At least four transcript variants encoding distinct isoforms have been found for this gene, but the full-length natures of only three of them have been determined. [provided by RefSeq]
Especificaciones
Especificaciones
| Antígeno | baculoviral IAP repeat-containing 5 |
| Aplicaciones | ELISA, Immunofluorescence, Immunoprecipitation, KnockDown, Western Blot |
| Clasificación | Monoclonal |
| Clon | 5B10 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against a partial recombinant BIRC5. |
| Formulación | PBS with no preservative; pH 7.4 |
| génica | BIRC5 |
| N.º de referencia del gen | NM_001168 |
| Alias de gen | API4/EPR-1 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.