Learn More
beaded filament structural protein 1, filensin, Mouse, Clone: 1B11, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant BFSP1.
Marca: Abnova H00000631-M01.100ug
Descripción
More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, CP49 (also known as phakinin) and the protein product of this gene, filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene are the cause of autosomal recessive cortical juvenile-onset cataract. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MYENECECQLLLKEMLERLNKEADEALLHNLRLQLEAQFLQDDISAAKDRHKKNLLEVQTYISILQQIIHTTPPASIVTSGMREEKLLTEREVAALRSQLEEGREVLSHLQAQRVELQAQTTTLEQAIKSAHECYDDEIQLYNEQIETLRKEIEETERVLEKSSYDCRQLAVAQQTLKNELDRYHRIIEIEGNRLTSAFIETPIPLFTQSHGVSLSTGSGGKDLTRALQDITAAKPRQKALPKNVPRRKEIITKDKTNGALEDAPLKGLEDTKLVQVVLKEESESKFESESKEVSPLTQEGAPEDVPDGGQISKGFGKLYRKVKEKVRSPKEPETPTELYTKERHVLVTGDANYVDPRFYVSSITAKGGVAVSVAEDSVLYDGQVGPSPESPKPPLENGQVGLQEKEDGQPIDQQPIDKEIEPDGAELEGPEEKREGEERDEESRRPCAMVTPGAEEPSIPEPPKPAADQDGAEVLGTRSRSLPEKGPPKALAYKTVEVVESIEKISTESIQTYEETAVIVETMIGKTKSDKKKSGEKSSEspecificaciones
| beaded filament structural protein 1, filensin | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC041483 | |
| BFSP1 | |
| BFSP1 (AAH41483, 1 a.a. ∼ 540 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 631 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| ELISA, Western Blot | |
| 1B11 | |
| Mouse monoclonal antibody raised against a full length recombinant BFSP1. | |
| BFSP1 | |
| CP115/CP94/FILENSIN/LIFL-H | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.