Learn More
Beclin Mouse, Unlabeled, Clone: 20, BD
Código de producto. 15836099
Change view
Click to view available options
Cantidad:
150 μg
50 μg
Tamaño de la unidad:
1 set
15836099 150 μg 1 set
15826099 50 μg 1 set
2 options
Código de producto. 15836099
Marca: BD Biosciences 612113
Mouse Monoclonal Antibody
Autophagy is the bulk degradation of cellular proteins through the autophagosomic-lysosomal pathway. This process is important for normal cell growth, and may be disrupted in tumor cells. Beclin is a Bcl-2 binding protein that is homologous to the yeast autophagy gene, apg6/vps30. The structure of beclin includes a Bcl-2 binding coiled-coil region, and a leucine-rich nuclear export signal (NES). Beclin protein colocalizes with intracytoplasmic organelles and nuclei in normal COS7 and MCF7 cells. However, inhibition of CRM1-dependent nuclear export leads to beclin localization primarily in the nucleus. Mutation of the NES domain of beclin also prevents nuclear export, as well as suppresses beclin-mediated nutrient deprivation-induced autophagy. In addition, these beclin mutants can not inhibit in vitro clonigenicity and in vivo tumorigenicity of MCF7 cells. Beclin interaction with Bcl-2 may be involved with host viral defense, since overexpression of beclin inhibits Sindbis virus replication and expression of beclin lacking the Bcl-2 binding domain has no antiviral effects. Thus, beclin may be an important component of complexes involved in autophagy.
Western Blotting
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Beclin
Aplicaciones Western Blot
Clasificación Monoclonal
Clon 20
Concentración 250μg/mL
Conjugado Unconjugated
Formulación Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Human Beclin aa. 171-291
Método de purificación Affinity Purified
Cantidad 150 μg
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Canine, Chicken, Human, Mouse, Rat
Contenido y almacenamiento Store undiluted at -20°C.
Isotype IgG2a
Mostrar más Mostrar menos
For Research Use Only.