Learn More
B7-H4 Mouse anti-Human, APC, Clone: MIH43, BD
Descripción
The MIH43 monoclonal antibody specifically binds to human B7-H4 (B7 family member, H4) that is encoded by the VTCN1 (V-set domain containing T cell activation inhibitor 1) gene. B7-H4 is a type I membrane glycoprotein with a calculated molecular weight of 30.89 kDa. It is also known as VCTN1, B7h.5, B7S1 (B7 superfamily member 1) or B7X. B7-H4 is a newly discovered member of the B7 family of costimulatory proteins. B7-H4 is not constitutively expressed on peripheral tissues. Its expression can be induced on macrophages, dendritic cells, B cells and T cells. By binding to its putative receptor, the function of B7-H4 was initially reported to be a negative regulator of T cell activation, proliferation and differentiation related to cytokine production and cytotoxic effector cell functions. B7-H4 is reportedly overexpressed in a variety tumors. B7-H4 expression by tumor macrophages appears to play a role in suppression antigen-specific T cell mediated immunity. Recent studies indicate that B7-H4 can also be a positive regulator of T cell responses. B7-H4 can be involved innate immunity as well, eg, by inhibiting neutrophil expansion.
Especificaciones
Especificaciones
| Antígeno | B7-H4 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | MIH43 |
| Conjugado | APC |
| Descripción | VTCN1; VCTN1; B7 family member, H4; B7H4; B7h.5; B7S1; B7X |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant Human B7-H4 protein |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.