Learn More
Annexin II Mouse, Unlabeled, Clone: 5, BD
Descripción
Annexin II (p36), originally discovered as a substrate for the src oncogene, is one of the most studied members of the annexin family of calcium-dependent phospholipid-binding proteins. It exists in cells as either a monomer or a heterotetramer complexed with p11, an S100-related protein. In vitro, it has been found that unphosphorylated annexin II is capable of causing aggregation of chromaffin granules, and, upon phosphorylation, induces fusion of the granules membranes. A similiar phenomenon may occur in the final steps of exocytosis after cytoplasmic annexin translocates to the subplasmalemmal region and is phosphorylated by protein kinase C. However, other studies refute annexin II's involvement in exocytosis, and a conclusive role for annexin II in membrane trafficking remains debated.
Host Species: Mouse
Clone: 5
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Annexin II aa. 123-328
Formula Weight [Chemical]: 36kDa
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Especificaciones
Especificaciones
| Antígeno | Annexin II |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 5 |
| Concentración | 250μg/mL |
| Conjugado | Unconjugated |
| Descripción | Lipocortin II, Calpactin I |
| Formulación | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human Annexin II aa. 123-328 |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.