Learn More
ALDH2, Mouse, Clone: 4F10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ALDH2.
Marca: Abnova H00000217-M10.100ug
Descripción
This protein belongs to the aldehyde dehydrogenase family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Orientals have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Orientals than among Caucasians could be related to the absence of the mitochondrial isozyme. This gene encodes a mitochondrial isoform, which has a low Km for acetaldehydes, and is localized in mitochondrial matrix. [provided by RefSeq
Sequence: DGMTIAKEEIFGPVMQILKFKTIEEVVGRANNSTYGLAAAVFTKDLDKANYLSQALQAGTVWVNCYDVFGAQSPFGGYKMSGSGRELGEYGLQAYTEVKTVTVKVPQKNSEspecificaciones
| ALDH2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC002967 | |
| ALDH2 | |
| ALDH2 (AAH02967, 408 a.a. ∼ 517 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 4F10 | |
| Mouse monoclonal antibody raised against a partial recombinant ALDH2. | |
| ALDH2 | |
| ALDH-E2/ALDHI/ALDM/MGC1806 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 217 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.