missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alcohol Dehydrogenase 1A Monoclonal Antibody (1E5)

Descripción
This antibody does not react with rabbit or rat protein.
This gene encodes a member of the alcohol dehydrogenase family. The encoded protein is the alpha subunit of class I alcohol dehydrogenase, which consists of several homo- and heterodimers of alpha, beta and gamma subunits. Alcohol dehydrogenases catalyze the oxidation of alcohols to aldehydes. This gene is active in the liver in early fetal life but only weakly active in adult liver. This gene is found in a cluster with six additional alcohol dehydrogenase genes, including those encoding the beta and gamma subunits, on the long arm of chromosome 4. Mutations in this gene may contribute to variation in certain personality traits and substance dependence.
Especificaciones
Especificaciones
| Antígeno | Alcohol Dehydrogenase 1A |
| Aplicaciones | Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 1E5 |
| Concentración | 2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% sodium azide; pH 7.4 |
| génica | ADH1A |
| N.º de referencia del gen | P07327 |
| Alias de gen | ADC1; Adh; ADH, alpha subunit; ADH1; Adh-1; ADH1A; Adh1c; Adh-1e; Adh1-e; Adh-1t; Adh-1-t; Adh1-t; Adh1tl; Adh-3e; Adh3-e; ADH-A2; ADH-AA; AI194826; Alcohol dehydrogenase 1; alcohol dehydrogenase 1 (class I); alcohol dehydrogenase 1 (class I), alpha polypeptide; alcohol dehydrogenase 1 temporal, liver; alcohol dehydrogenase 1, complex; alcohol dehydrogenase 1, electrophoretic; alcohol dehydrogenase 1A; alcohol dehydrogenase 1A (class I), alpha polypeptide; alcohol dehydrogenase 3, electrophoretic; alcohol dehydrogenase A subunit; alcohol dehydrogenase ADH1; Alcohol dehydrogenase I; Alcohol dehydrogenase subunit alpha; aldehyde reductase; class I alcohol dehydrogenase; O0947; YADH-1; YOL086C |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?