Learn More
AIP1 Mouse, Unlabeled, Clone: 49, BD
Descripción
Apoptosis is a selective process of genetically programmed cell death which occurs during normal cell differentiation and development of multicellular organisms. In vertebrates, T cell and neuronal development are probably the best characterized systems for the study of apoptosis. ALG-2 and ALG-3 (apoptosis-linked genes 2 and 3) were identified as low molecular weight Ca2+ -binding proteins essential for apoptosis through the activation of the Fas receptor in T cells. ALG-2 Interacting Protein 1 (AIP1/Alix) is a ubiquitous protein that associates with ALG-2 in the cytosol in a Ca2+; dependent manner. AIP1 is homologous to the yeast protein, BRO1, which has been implicated in Pkc1p- AP kinase signaling. A truncated form of AIP1 protects against serum starvation-, etoposide-, and staurosporine-induced cell death. In addition, the C-terminal proline rich region of AIP1 facilitates interaction with SH3 domain-containing protein expressed in tumorigenic astrocytes (SETA) and this interaction may be important for mediating DNA damage-dependent apoptosis in astrocytes. Thus, AIP1 interacts with ALG-2 or SETA, or both, during activation of cell death pathways in a variety of cell types.
Host Species: Mouse
Clone: 49
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Mouse AIP1 aa. 375-580
Immunofluorescence, Western Blotting
Especificaciones
Especificaciones
| Antígeno | AIP1 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 49 |
| Concentración | 250μg/mL |
| Conjugado | Unconjugated |
| Descripción | ALG-2 Interacting Protein 1; Alix |
| Formulación | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Mouse AIP1 aa. 375-580 |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.