missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF-1/Iba1 Goat anti-Human, Mouse, Rat, Canine, Feline, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody has been used in 192 publications
Marca: Novus Biologicals NB100-1028
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
AIF-1/Iba1 Polyclonal specifically detects AIF-1/Iba1 in Human, Mouse, Rat, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockout Validated, Dual RNAscope ISH-IHC.Especificaciones
| AIF-1/Iba1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1 | |
| Goat | |
| 16.7 kDa | |
| 0.1 mg | |
| Primary | |
| This AIF-1/Iba1 Antibody is expected to recognize isoform 1 (NP_116573.1) and isoform 3 (NP_001614.3). AIF-1/Iba1 is thought to be involved in negative regulation of growth of vascular smooth muscle cells, which contributes to the anti-inflammatory response to vessel wall trauma. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ELISA, Immunohistochemistry - Free Floating, Immunoassay | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1.0 ug/mL, Immunohistochemistry 4 - 6 ug/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 4 - 6 ug/mL, Immunohistochemistry-Frozen, Peptide ELISA Detection Limit 1:64000, Immunohistochemistry Free-Floating, Immunohistochemistry, Knockout Validated, Dual RNAscope ISH-IHC | |
| P55008 | |
| AIF1 | |
| This AIF-1/Iba1 Antibody was developed against a peptide with sequence C-TGPPAKKAISELP, from the C Terminus of the protein sequence according to NP_116573.1; NP_001614.3. | |
| Affinity Purified | |
| RUO | |
| 199 | |
| Human, Mouse, Rat, Canine, Feline | |
| IgG |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
For Research Use Only