Learn More
ADAM metallopeptidase with thrombospondin type 1 motif, 13, Mouse, Polyclonal Antibody, Abnova™
Descripción
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene is the von Willebrand Factor (vWF)-cleaving protease, which is responsible for cleaving at the site of Tyr842-Met843 of the vWF molecule. A deficiency of this enzyme is associated with thrombotic thrombocytopenic purpura. Alternative splicing of this gene generates multiple transcript variants encoding different isoforms. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | ADAM metallopeptidase with thrombospondin type 1 motif, 13 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Mouse polyclonal antibody raised against a partial recombinant ADAMTS13. |
| Formulación | 50% glycerol |
| génica | ADAMTS13 |
| N.º de referencia del gen | NM_139025 |
| Alias de gen | C9orf8/DKFZp434C2322/FLJ42993/MGC118899/MGC118900/TTP/VWFCP/vWF-CP |
| Símbolos de los genes | ADAMTS13 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.