Learn More
anti-active/cleaved Caspase 3, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 48 publications
Marca: Novus Biologicals NB100-56113
Descripción
Caspase-3 Polyclonal specifically detects Caspase-3 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular).Especificaciones
| Caspase-3 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular) | |
| Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
| Rabbit | |
| 31.7 kDa | |
| 0.05 mL | |
| Primary | |
| Preferentially detects active caspase-3 (large subunit: ∼14-21 kDa, and small subunit: ∼10 kDa). However, it may also detect pro-caspase-3 (∼32 kDa) in some cell or tissue systems. Nuclear immunostaining of caspase-3 is considered be an indication of active/cleaved caspase-3. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
| Unconjugated | |
| Neat whole antisera with 0.05% Sodium Azide | |
| CASP3 | |
| This Caspase-3 Antibody - (active/cleaved) was developed against catalytically active human caspase-3 protein. | |
| Unpurified | |
| RUO | |
| 836 | |
| Human, Mouse, Rat, Gerbil | |
| Antisera |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
For Research Use Only