Learn More
ABHD1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ABHD1 protein.
Marca: Abnova H00084696-B01P.50ug
Descripción
This gene is a member of the AB hydrolase superfamily and encodes a protein with an alpha/beta hydrolase fold. This domain is common to a number of hydrolytic enzymes of widely differing phylogenetic origins and catalytic functions. [provided by RefSeq
Sequence: MLSSFLSPQNGTWADTFSLLLALAVALYLGYYWACVLQRPRLVAGPQFLAFLEPHCSITTETFYPTLWCFEGRLQSIFQVLLQSQPLVLYQSDILQTPDGGQLLLDWAKQPDSSQDPDPTTQPIVLLLPGITGSSQETYVLHLVNQALRDGYQAVVFNNRGCRGEELRTHRAFCASNTEDLETVVNHIKHRYPQAPLLAVGISFGGILVLNHLAQARQAAGLVAALTLSACWDSFETTRSLETPLNSLLFNQPLTAGLCQLVERNRKVIEKVVDIDFVLQARTIRQFDERYTSVAFGYQDCVTYYKAASPRTKIDAIRIPVLYLSAADDPFSPVCALPIQAAQHSPYVALLITARGGHIGFLEGLLPWQHWYMSRLLHQYAKAIFQDPEGLPDLRALLPSEDRNSEspecificaciones
| ABHD1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ABHD1 protein. | |
| ABHD1 | |
| FLJ36128/LABH1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84696 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC039576.1 | |
| ABHD1 | |
| ABHD1 (AAH39576.2, 1 a.a. ∼ 405 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.