missing translation for 'onlineSavingsMsg'
Learn More
Learn More
53BP1 Rabbit anti-Human, Mouse, Other, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 96 publications
Marca: Novus Biologicals NB100-904
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
53BP1 Polyclonal specifically detects 53BP1 in Human, Mouse, Other samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Proximity Ligation Assay, In-situ Hybridization, Knockdown Validated.Especificaciones
| 53BP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7158 | |
| Human, Mouse, Other | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), In situ PLA, In Situ Hybridization (ISH), KnockDown | |
| 1 mg/mL | |
| Western Blot 1:1000 - 1:10000, Flow Cytometry 2 - 5 μg/mL, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:400, Proximity Ligation Assay, In-situ Hybridization, KnockDown Validated | |
| Q12888 | |
| TP53BP1 | |
| 53BP1 Antibody was made to a synthetic peptide corresponding to a portion of human 53BP1 encoded in exon 11, 12 and 19 [Uniprot: Q12888]. This antibody is a cocktail hybrid of NB100-304 and NB100-305. | |
| 0.1 mL | |
| Primary | |
| Human and mouse. Human has been tested in WB, IHC and ICC/IF, mouse has only been tested in ICC/IF. | |
| Store at -20 degreesC. |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
For Research Use Only