missing translation for 'onlineSavingsMsg'
Learn More

53BP1 Mouse anti-Human, Unlabeled, Clone: 19, BD

Código de producto. 15806209
Change view
Click to view available options
Cantidad:
150 μg
50 μg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15806209 50 μg cada uno
15816209 150 μg cada uno
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15806209

Marca: BD Biosciences 612522

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The p53 protein is critical to regulation of normal cell growth and is a suppressor of tumor cell proliferation. Inactivation of p53 by a number of mechanisms, such as missense mutations or interaction with oncogenic viral or cellular proteins, can result in tumor progression. In addition, Bcl2 and p53 are involved in apoptosis in an antagonistic fashion such that overexpressed Bcl2 inhibits p53-induced apoptosis. 53BP1 and 53BP2 were identified in a yeast two-hybrid screen of proteins that bind p53. Both 53BP1 and 53BP2 bind wild type p53, but not mutant p53 found in tumor cells. p53BP1 is localized to the cytoplasm and nucleus, while p53BP2 is found only in the cytoplasm. 53BP1 has BRCT motifs found in proteins involved in cell cycle control and DNA repair. DNA damage leads to 53BP1 hyperphosphorylation, which may be mediated by ATM. 53BP2 has four ankyrin repeats and a SH3 domain that are required for interactions with Bcl2 and p53. Overexpression of 53BP2 in 293 cells inhibits progression of the cell cycle in G2/M phase, while co-transfection of 53BP2 with p53 in H358 cells enhances p53-mediated transcriptional activation. The interaction between 53BP2 and p53 may be regulated by Bcl2, since competition experiments demonstrate that Bcl2 prevents p53 binding to 53BP2. In addition, 53BP2 can also bind the apoptotic-related p65 subunit of NFκB and this subunit can inhibit 53BP2-induced cell death.

Bioimaging, Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno 53BP1
Aplicaciones Western Blot
Clasificación Monoclonal
Clon 19
Concentración 250μg/mL
Conjugado Unconjugated
Formulación Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Human 53BP1 aa. 149-259
Método de purificación Affinity Purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at -20°C.
Isotype IgG2b
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.