Descripción
- Sterile glove made from a synthetic, nitrile formulation
- High elasticity with barrier protection and puncture resistance
- Ideal solution for a range of sterile examination (non-surgical) procedures
- Packed as a pair of gloves per pack
- 12 in. extended cuff for additional protection
- Textured fingers provide extra grip and reliability
- Gamma sterilized minimum 25kGγ to provide sterility
- Tested for use with chemotherapy drugs
Especificaciones
Especificaciones
| Certificaciones/conformidad | ASTM D6978, ASTM D6319, , ISO 13485, ISO 9001, ISO 10993-10:2002 |
| Certificaciones/conformidad (Europa) | EN ISO 374-5:2016, EN ISO 374-1:2016, EN 420:2003 + A1:2009 |
| Aplicaciones | Asistencia sanitaria |
| AQL | 1.5 |
| Color | Azul |
| Desechables | Single-use |
| Material | Nitrilo |
| Tamaño | Pequeño |
| Modelo | 6034151 |
| Longitud (imperial) | 11,6 in |
| Mostrar más |