Learn More
STEMCELL Technologies Annexin V Apoptosis Detection Kit with 7-AAD, FITC
Descripción
- The Annexin V Apoptosis Detection Kit includes Annexin V (Catalog #100-0329 or Catalog #100-0331 or Catalog #100-0333), Annexin V Binding Buffer (Catalog #100-0334), and 7-AAD (Catalog #75001)
- The kit is used for the combined detection of early-stage cell apoptosis (using Annexin V) and late-stage cell apoptosis or necrosis (using both Annexin V and 7-AAD)
- Annexin V belongs to the annexin family of intracellular proteins that bind to membrane phospholipids in a calcium-dependent manner
- This dye has a high affinity for phosphatidylserine (PS) that is present in the inner leaflet of the plasma membrane
- During early-stage cell apoptosis, PS is translocated from the inner to the outer leaflet of the cell membrane, exposing it to the external environment, where it is detected by Annexin V
- This process of PS translocation occurs prior to the loss of membrane integrity
- As cells progress through apoptosis and towards necrosis, the cell membrane is compromised and consequently the 7-AAD viability dye passes into the cell
- Thus, cells undergoing early apoptosis stain positive for Annexin V and negative for viability dyes, while apoptotic death or necrosis is characterized by positive staining for both Annexin V and 7-AAD
Especificaciones
Especificaciones
| Cantidad | 100 pruebas |
| Para utilizar con (aplicación) | Flow Cytometry |
| Conjugado | FITC |