Learn More
Invitrogen™ Annexin A7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA592049
Descripción
Immunogen sequence: TYYDAWSLRK AMQGAGTQER VLIEILCTRT NQEIREIVRC YQSEFGRDLE KDIRSDTSGH FERLLVSMCQ GNRDENQSIN HQMAQEDAQR LYQAGEGRLG TDESCFNMIL ATRSFPQLRA TMEAYSRMAN RDLLSSVSRE FSGYVESGLK TILQCALNRP AFFAERLYYA MKGAGTDDST LVRIVVTRSE IDLVQIKQMF AQMYQKTLGT MIAGDTSGDY RRLLLAIVGQ; Positive Samples: SH-SY5Y, NCI-H460, HepG2, SW480, NIH/3T3, Mouse heart, Mouse lung, Rat lung.
Calcium/phospholipid-binding protein which promotes membrane fusion and is involved in exocytosis. Annexin VII is a member of the annexin family of calcium-dependent phospholipid binding proteins. The Annexin VII gene contains 14 exons and spans approxima.
Especificaciones
| Annexin A7 | |
| Polyclonal | |
| Unconjugated | |
| ANXA7 | |
| AI265384; AI316497; Annexin A7; annexin VII; Annexin-7; Anx7; ANXA7; Interleukin-8; OK/SW-cl.95; RP11-537A6.8; SNX; SYNEXIN | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11750, 155423, 310 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.04 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P20073, Q07076 | |
| ANXA7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 237-466 of human ANXA7 (NP_0011471). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.