Descripción
The Annexin A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Annexin A2. This antibody recognizes human, mouse antigen. The Annexin A2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Especificaciones
Especificaciones
| Antígeno | Annexin A2 |
| Aplicaciones | Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | 3H1B11 |
| Conjugado | CoraLite 594 |
| Especie del huésped | Mouse |
| Cantidad | 100 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 302 |
| Especies diana | Human, Mouse |
| Mostrar más |