missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A1 Antibody (ANXA1/3566), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-08708DL594
This item is not returnable.
View return policy
Description
Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Annexin A1 | |
| Monoclonal | |
| DyLight 594 | |
| annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
| Recombinant full-length human Annexin A1 protein (Uniprot: P04083) | |
| 100 μg | |
| Protein Kinase, Signal Transduction | |
| 301 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| ANXA1/3566 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction