missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKIB1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NB100-61607AF594
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ANKIB1 Polyclonal antibody specifically detects ANKIB1 in Human samples. It is validated for Western Blot
Especificaciones
| ANKIB1 | |
| Polyclonal | |
| Western Blot | |
| ankyrin repeat and IBR domain containing 1, ankyrin repeat and IBR domain-containing protein 1, DKFZp434A0225, FLJ33123, KIAA1386DKFZP434A0225 | |
| The immunogen recognized by this antibody maps to a region between residue 925 and 975 of human ankyrin repeat and IBR domain-containing protein 1 using the numbering given in entry NP_061877.1 (GeneID 54467). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 54467 | |
| Store at 4C in the dark. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Spot an opportunity for improvement?Share a Content Correction