missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANAPC4, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Descripción
Sequence: VTVVLKDTVGREGRDRLLVQLPLSLVYNSEDSAEYQFTGTYSTRLDEQCSAIPTRTMHFEKHWRLLESMKAQYVAGNGFRKVSCVLSSNLRHV
Especificaciones
Especificaciones
| Antígeno | ANAPC4 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:20-1:50) The optimal working dilution should be determined by the end user. |
| Formulación | In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). |
| génica | anaphase promoting complex subunit 4 |
| Alias de gen | APC4 |
| Símbolos de los genes | ANAPC4 |
| Especie del huésped | Rabbit |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?