missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Ampicillin trihydrate is a β-lactam antibiotic with an amino group side chain attached to the penicillin structure. It is a penicillin derivative that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane.
- Inactivated by beta-lactamases; therefore, a beta-lactamase inhibitor should be considered when using ampicillin
- Antimicrobial spectrum: gram-negative and gram-positive bacteria.
- Against gram-positive bacteria, ampicillin has a similar mode of action as benzylpenicillin
- Against gram-negative bacteria, it has a similar mode of action as chloramphenicol and tetracyclines
- Bactericidal only to growing E. coli. Inhibits cell wall synthesis via cleavage of β-lactam ring of ampicillin by β-lactamase
Especificaciones
Especificaciones
| Forma física | Powder |
| Punto de fusión | 198°C to 200°C (literature) |
| Concentración | Working Concentration: 100mg/liter for both gram positive and gram negative bacteria. It is typically stable in media at 37°C for approximately 3 days. |
| Cantidad | 5 g |
| Información de solubilidad | Soluble in 1M Ammonium Hydroxide (50mg/mL), dilute acids (50mg/mL 1M HCl) or bases; slightly soluble in water (10mg/mL); practically insoluble in alcohol, chloroform, ether and fixed oils. |
| Formula Weight (peso de la fórmula) | 403.46 |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?