missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOKU-E™ Ampicillin ReadyMade™ Solution
Descripción
- CAS Number: 69-53-4
- Molecular Formula: C16H19N3O4S (ampicillin anhydrous)
- Molecular Weight: 349.40 g/mol
- Storage Conditions: -20°C
- Form: Liquid
- Source: Semi-synthetic
- Spectrum: Ampicillin is broad-spectrum, effective against both Gram-positive and Gram-negative bacteria including Gram-negative non-extended spectrum β-lactamase (ESBL) bacteria including Staphylococcus and Streptococcus species and medically important enteric pathogens such as Shigella and Salmonella. Interestingly, Ampicillin has been found to be effective against certain β-lactam sensitive vancomycin-resistant Enterococcus (VRE); a glycopeptide antibiotic resistant 'superbug'. Resistance to Ampicillin is routinely used as a selectable marker to confirm successful cell transformation
- Mechanism of Action: Like all β-lactams, Ampicillin interferes with penicillin binding protein (PBP) activity, otherwise involved in the final phase of peptidoglycan synthesis. PBPs are enzymes which catalyze a pentaglycine crosslink between alanine and lysine residues. Without a pentaglycine crosslink, the integrity of the cell wall is severely compromised ultimately leading to cell lysis
Especificaciones
Especificaciones
| CAS | 69-53-4 |
| Certificaciones/conformidad | Purity >98.0% |
| Nombre del producto químico o material | Ampicillin |
| Color | Opaque white to opaque yellow |
| Contenido y almacenamiento | -20°C |
| Descripción | Ampicillin Readymade™ Solution 100 mg/mL in DMSO, Sterile Filtered |
| Concentración | 100 to 105 mg/mL |
| Formulario | Líquido |
| Tipo de producto | Antibiotic |
| Esterilidad | Estéril |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?