Descripción
- It is a widely used reagent in molecular biology and chromatography.
- It is useful in the isolation and purification of enzymes and proteins.
- It is used for the precipitation or fractionation of proteins or for purification of antibodies.
- It is also useful for crystallographic analysis of nucleic acids and proteins.
- It is also widely used in HPLC of proteins, such as in hydrophobic interaction chromatography.
- It is used in long PCR buffer, in PCR lysis solution, and in secondstrand cDNA synthesis.
- it is used for the HPLC analysis of phospholipids.
- Ammonium sulfate is an inorganic salt prepared by treating ammonia with sulfuric acid.
- Ammonium sulfate decomposes upon heating above 250°C, forming ammonium bisulfate.
- Heating at higher temperatures results in decomposition into ammonia, nitrogen, sulfur dioxide, and water.
- Ammonium sulfate forms many double salts (ammonium metal sulfates) when its solution is mixed with equimolar solutions of metal sulfates and the solution is slowly evaporated.
- Such double metal sulfates include ammonium cobaltous sulfate, ferric ammonium sulfate, ammonium nickel sulfate and ammonium cerous sulfat
Especificaciones
Especificaciones
| Punto de fusión | >280°C (decomposition)(literature) |
| Color | Blanco |
| pH | 5 to 6 (5% aq soln) |
| Forma física | Crystalline Powder |
| Cantidad | 50 g |
| Notas de grado de pureza | Ultra Puregrade |
| Información de solubilidad | Soluble freely in water; insoluble in alcohol, acetone. |
| Porcentaje de pureza | ≥99% |
| Residuo después del encendido | 8e-006 |
| Densidad | 1.77g/mL at 25°C (Lit.) |
Seguridad y manipulación
Nombre del producto
- Ammonium Sulphate, ultra pure
- PELIGRO ACUÁTICO A LARGO PLAZO Categoría 4
- H413-Puede ser nocivo para los organismos acuáticos, con efectos nocivos duraderos.
- P101-Si se necesita consejo médico, tener a mano el envase o la etiqueta.
- P102-Mantener fuera del alcance de los niños.
- P103-Leer la etiqueta antes del uso.
- P273-Evitar su liberación al medio ambiente.
- P501b-Eliminar el contenido/el recipiente en conforme a la reglamentación local/regional/nacional/internacional.
- MIXTURE LIST-Contém: Ammonium Sulphate