Learn More
Sulfato de amonio y hierro(III) dodecahidrato, 98,5-102,0 %, Thermo Scientific Chemicals
Descripción
Ammonium iron(III) sulfate dodecahydrate is used in dyeing and printing textiles, as analytical lab reagent. It is often employed as an analytical standard, and has been used in a variety of other applications from nanomaterials to general redox reactions. It plays an important role for the inactivation of mycobacteriophage D29 bacteria which is vivale for viable mycobacterium smegmatis and m.tuberculosis. The crystalline ammonium ferrous sulfate has shown useful function as the high absorbed dose and low-temperature dosimeter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Ammonium iron(III) sulfate dodecahydrate |
| Punto de fusión | 39°C to 41°C |
| Nombre de nota | For analysis ACS |
| Color | Morado |
| Forma física | Cristales rotos |
| Cantidad | 2.5 kg |
| Intervalo de porcentaje de ensayo | 98.5-102.0% |
| Información de solubilidad | Freely soluble in water and hydrochloric acid. Insoluble in alcohol. |
| Formula Weight (peso de la fórmula) | 482.18 (266.00 Anhydrous) |
| Grado | ACS |
| Mostrar más |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.