missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AM679, MedChemExpress
Descripción
AM679 is a potent, selective 5-lipoxygenase-activating protein (FLAP) inhibitor with an IC50 of 2 nM in a human FLAP membrane binding assay. AM679 markedly reduces the respiratory syncytial virus-driven ocular pathology as well as the synthesis of cysteinyl leukotrienes (CysLTs) in the eye.
Especificaciones
Especificaciones
| Nombre del producto químico o material | AM679 |
| CAS | 1206880-66-1 |
| Color | pale pink |
| Notas de grado de pureza | Research |
| Fórmula molecular | C40H44N4O5S |
| Cantidad | 50 mg |
| Información de solubilidad | DMSO : ≥ 100 mg/mL (144.33 mM) |
| SMILES | O=C(C)N1C2=CC=CC=C2C[C@H]1COC3=CC=C(N(CC4=CC=C(C5=NC=C(OC)C=N5)C=C4)C(CC(C)(C(O)=O)C)=C6SC(C)(C)C)C6=C3 |
| Peso molecular (g/mol) | 692.87 |
| Formula Weight (peso de la fórmula) | 692.87 |
| Mostrar más |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?