missing translation for 'onlineSavingsMsg'
Learn More

AM-2099, MedChemExpress

Product Code. 16474426
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Unit Size:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
Product Code. Cantidad unitSize
16474426 100 mg 100 miligramos
16489686 50 mg 50 miligramos
16485867 5 mg 5 miligramos
16443138 10 mg 10 miligramos
4 options
This item is not returnable. View return policy

Product Code. 16474426

Brand: Medchem Express HY100727

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress AM-2099 is a potent and selective inhibitor of voltage-gated sodium channel Nav1.7 with an IC50 of 0.16 μM for human Nav1.7.

AM-2099 is a potent and selective inhibitor of voltage-gated sodium channel Nav1.7 with an IC50 of 0.16 μM for human Nav1.7.

Specifications

Nombre del producto químico o material AM-2099
CAS 1443373-17-8
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C19H13F3N4O3S2
Cantidad 100 mg
Información de solubilidad DMSO : ≥ 150 mg/mL (321.57 mM)
SMILES COC1=CC(C(F)(F)F)=CC=C1C2=NC=NC3=CC(S(=O)(NC4=NC=CS4)=O)=CC=C32
Peso molecular (g/mol) 466.46
Formula Weight (peso de la fórmula) 466.46
Porcentaje de pureza 98.02%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.