Descripción
- Specialized protection:The AlphaTec™ 4000 Ultrasonically Welded and Taped - Model 130 is a robust chemical protective suit, permeation-tested to withstand more than 200 hazardous chemicals, including chemical warfare agents
- Increased strength: This full body protection suit comes with taped and welded seams to grant users added strength and chemical barrier defense
- Heightened visibility: Designed with orange-colored material, it ensures better visibility which can reduce the risk of workplace injuries
- Improved practicality: It also possesses a chinstrap and a thumb loop, which are compatible with a range of respirators and hand protection products
- Optimized fit: Its elasticated two-piece hood, waist, cuffs and ankles simplify the donning and doffing process
- Certified anti-static capabilities: In addition, this anti-static suit complies with EN 1149-5
- CE Category III Type 3/4/5 protection
Especificaciones
Especificaciones
| Color | Naranja |
| Hood Type | Attached |
| Material | Multi-layer Barrier Laminate Fabric |
| Tamaño | Small |
| Estilo de tobillo | Elasticated |
| Tipo de tapón | Single zip closure |
| Tipo de collarín | 3-piece hood |
| Estilo de manguito | Elasticated |
| Resistencia | Chemical |
| Tipo de costura | Ultrasonicaly welded and taped |