missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
- Specialized protection:The AlphaTec™ 4000 Ultrasonically Welded and Taped - Model 130 is a robust chemical protective suit, permeation-tested to withstand more than 200 hazardous chemicals, including chemical warfare agents
- Increased strength: This full body protection suit comes with taped and welded seams to grant users added strength and chemical barrier defense
- Heightened visibility: Designed with orange-colored material, it ensures better visibility which can reduce the risk of workplace injuries
- Improved practicality: It also possesses a chinstrap and a thumb loop, which are compatible with a range of respirators and hand protection products
- Optimized fit: Its elasticated two-piece hood, waist, cuffs and ankles simplify the donning and doffing process
- Certified anti-static capabilities: In addition, this anti-static suit complies with EN 1149-5
- CE Category III Type 3/4/5 protection
Especificaciones
Especificaciones
| Color | Naranja |
| Hood Type | Attached |
| Material | Multi-layer Barrier Laminate Fabric |
| Tamaño | 2X-Large |
| Estilo de tobillo | Elasticated |
| Tipo de tapón | Single zip closure |
| Tipo de collarín | 3-piece hood |
| Estilo de manguito | Elasticated |
| Resistencia | Chemical |
| Tipo de costura | Ultrasonicaly welded and taped |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?