Learn More
Ansell AlphaTec™ 3000 Ultrasonically Welded Model 122
Tienda Ansell Productos
Click to view available options
Tamaño:
4X-Large
5X-Large
Tamaño de la unidad:
cada uno
Descripción
- Specialized protection: Made with durable multi-layer nonwoven laminate, the AlphaTec™ 3000 Ultrasonically Welded, Model 122 protects against various inorganic chemicals and biological hazards, while ensuring lightweight comfort
- Enhanced features: This chemical-protective suit features dual-zip systems, double cuffs and attached barrier gloves, enhancing its protective capabilities, as well as a two-piece hood and an elasticated waist, sleeves and over-cuffs
- Improved visibility: Thanks to its high-visibility yellow exterior, this protective suit ensures greater wearer and workplace safety
- Assured antistatic capabilities: This full-body protection suit is also tested against EN 1149-5 antistatic standards
Especificaciones
Especificaciones
| Tipo de producto | Ropa de protección química |
| Tamaño | 4X-Large |
| Sexo | Unisex |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Desechables | Reusable |
| Estilo de cintura | Elástico |
| Estilo de tobillo | Attached Socks, Boot Overflaps |
| Estilo de manguito | Elastic |
| Estilo de cuello | Hooded |
| Mostrar más |