Learn More
Ansell AlphaTec™ 3000 Avant2 Airline Ultrasonically Welded and Taped - Model 757
Código de producto. 17161105
Change view
Click to view available options
Tamaño:
Small
Medium
Grande
X-Large
2X-Large
3X-Large
Tamaño de la unidad:
paquete de 4
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Tamaño unitSize
17161105 3X-Large paquete de 4
17111095 Small paquete de 4
17121095 Small paquete de 4
17141095 Medium paquete de 4
17151095 Medium paquete de 4
17171095 Grande paquete de 4
17181095 Grande paquete de 4
17101105 X-Large paquete de 4
17111105 X-Large paquete de 4
17131105 2X-Large paquete de 4
17141105 2X-Large paquete de 4
17171105 3X-Large paquete de 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Código de producto. 17161105 Proveedor Ansell N.º de proveedor YE30TA0075707
Solo quedan null
Añadir a la cesta
Añadir a la cesta
AlphaTec™ 3000 Avant2 Airline Ultrasonically Welded and Taped Chemical Protective Suit - Model 757 with HEPA filter offers secondary protection from airline contamination and a SMC silencer maintains the noise within a suit below 70 dB (even at maximum airflow).
  • Specialized protection: The AlphaTec™ 3000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 757 full body protection suit has been tested against 200 chemicals.
  • Genuine versatility: This chemical protective suit's HEPA filter offers secondary protection from airline contamination and a SMC silencer maintains the noise within a suit below 70 dB (even at maximum airflow)
  • Considered aesthetics: In addition, this protective suit has attached socks with an elasticated boot overflap and is designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress
  • Reduced risks: The emergency ripcord feature of this full protective suit permits quick doffing of the suit in cases of emergency or undue distress to the wearer
DESIGN FEATURES:
  • Fully encapsulated suit designed for use in combination with the permanently attached AlphaTec™ AVANT STS V.1 airline regulator.
  • Suit features include internal air-channel ventilation system double cuffs, finger and thumb loop, G01 Ansell Solvex (Nitrile) gloves, G02 Ansell Barrier gloves or G05 AlphaTec™ 29-500 gloves.
  • Attached socks with elasticated boot over-flap and emergency rip cord.
TRUSTED_SUSTAINABILITY
Especificaciones
Certificaciones/conformidad CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Color Yellow
Para utilizar con (aplicación) Opening and draining pumps valves or lines, Filling, blending and charging of raw materials, Contaminated Environments, Chemical Handling, Transferring liquids and solids, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes, Loading and unloading of proces equipment
Hood Type Totalmente encapsulado
Material Multi-layer Barrier Laminate Fabric
Tamaño 3X-Large
Estilo de tobillo Attached Socks, Elasticated Boot Over Flap
Desechables Reusable
Para utilizar con (equipo) Air-Fed/Supplied Systems
Sexo Unisex
Estilo de cuello Hood
Tipo de producto Encapsulated Chemical Protective Suit
Clase de protección CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Tipo de costura Soldada y con cinta
Esterilidad No estéril