Learn More
Ansell Alphatec™ 3000 Apron with Sleeves Ultrasonically Welded - Model 243-G01
Description
DESIGN FEATURES:
- Apron with sleeves with rear velcro fastening at the neck and attached AlphaTec™ 37-676
- Unexpected leakages spills or other releases
- Loading and unloading trucks and vehicles
- Loading trucks and vehicles
- Product inspection
- Cleaning of plant and machines
Specifications
Specifications
| Tipo | Chemical-resistant Apron |
| Tamaño | Small |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Desechables | Reusable |
| Tipo de tapón | Hook and Loop |
| Esterilidad | No estéril |
| Peso (métrico) | 80 gsm |
| Tipo de costura | Welded |
| Certificaciones/conformidad (Europa) | CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, EN 14126) |