Descripción
- Specialized protection: The AlphaTec™ 1500 PLUS Stitched - Model 103 protective suit is made from SMS material, protecting wearers against liquid spray and particulates, filtering 99.9% of particulates over 3 microns
- Heightened breathability: Air and water vapor permeability help this protective outer garment minimize the risk of excessive heat stress and maximize wearer comfort
- Assured antistatic capabilities: The AlphaTec™ 1500 PLUS Stitched - Model 103 is also tested against EN 1149-5 and AATC 76 antistatic standards
- Enhanced fit and features: Its optimized fit ensures comfort, with additional features including a two-way zipper with resealable storm flap, finger loops and an elasticated waist, wrist and ankles
Especificaciones
Especificaciones
| Tipo de producto | Collared Coveralls |
| Tamaño | 3XL |
| Sexo | Unisex |
| Material | SMS antiestática |
| Color | Blanco |
| Desechables | Reusable |
| Estilo de cintura | Elastic at Back |
| Estilo de tobillo | Elastic |
| Estilo de manguito | Finger Loops |
| Estilo de cuello | Con anillo |
| Mostrar más |