Learn More
Ansell Alphatec™ 1500 Plus Lab Coat Bound - Model 204
Tienda Ansell Productos
Click to view available options
Tamaño:
Medium
Grande
X-Large
2X-Large
Tamaño de la unidad:
paquete de 60
Descripción
- Optimized breathability: AlphaTec™ 1500 PLUS Lab Coat Bound - Model 204 delivers comfortable protection in general industrial environments, thanks to its air and water vapor permeable as well as anti-static, SMS material
- Reduced contamination risks: As this protective outer garment is silicone-free, it minimizes contamination risks, making it ideal for the life sciences and metal fabrication industries
- Improved ergonomics: In addition, this white protective over garment offers an optimized fit, which improves wearer comfort and workplace safety
DESIGN FEATURES:
- Medium length labcoat with collar, studded front fastening, left breast and right thigh pockets.
Especificaciones
Especificaciones
| Tipo de producto | Anti-static Lab Coat |
| Tamaño | X-Large |
| Sexo | Unisex |
| Material | SMS |
| Color | Blanco |
| Tipo de tapón | Botones |
| Estilo de manguito | Elástico |
| Desechables | Reusable |
| Esterilidad | No estéril |
| Resistencia | Chemicals |
| Mostrar más |