Learn More
alpha-synuclein Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16897493 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16897493 20 μL 20 microlitros
16887493 150 μL 150 microlitros
2 options
Código de producto. 16897493
Marca: Proteintech 108421AP20UL
Rabbit Polyclonal Antibody
This antibody can recognize all the isoforms of alpha-syn.
Alpha-Synuclein belongs to the Synuclein family, which includes beta and gamma Synuclein, and is predomitly expressed in neurons, and concentrated at synaptic terminals. Synucleins may be involved in the regulation of dopamine release and transport and the modulation of synaptic vesicle function. Alpha-Synuclein is a small, presynaptic protein that is the major nonamyloid component of the pathological inclusions characteristic of a wide range of neurodegenerative disorders, collectively known as synucleinopathies. Alpha-Synuclein is involved in the formation of SNARE complexes, and most significantly, aggregated alpha-synuclein is one of the major components found in the Lewy bodies that occur in Parkinson′s disease (PD) and other neurodegenerative disorders. Mutation of the alpha-synuclein gene is associated with familial forms of PD. Alpha-Synuclein is known to reduce the fibrillization of the microtubule-associated protein, tau, to bind various targets such as 14-3-3, protein kinase C, synphilin-1, Elk and Tat-binding protein 1. The alpha and beta synucleins are found primarily in brain tissue where they are seen mainly in neuronal cytosol and presynaptic terminals. The antibody specific for alpha-synuclein does not cross react with beta-synuclein and will be useful for the confirmation of Parkinson′s disease and Lewy body dementia and other studies for the localization and detection of alpha-synuclein. Four alternatively spliced transcripts that encode two different isoforms of the alpha-synuclein protein have been identified.
Especificaciones
Antígeno alpha-synuclein
Aplicaciones Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.51 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica SNCA
N.º de referencia del gen O55042, P37377, P37840
Alias de gen Alpha synuclein, NACP, PARK1, PARK4, SNCA
Símbolos de los genes SNCA
Especie del huésped Rabbit
Inmunógeno alpha-synuclein Fusion Protein Ag1285
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 20617, 29219, 6622
Especies diana Mouse, Human, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG