Learn More
Alpha SMA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16886595 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16886595 150 μL 150 microlitros
16896595 20 μL 20 microlitros
2 options
Código de producto. 16886595
Marca: Proteintech 551351AP150UL
Rabbit Polyclonal Antibody
This antibody stains the aorta only.
Among the six actin isoforms described in mammals, two are found in virtually all cells (b- and g-cytoplasmic), two are detected in smooth muscle cells (a- and g-smooth muscle) and two are present in striated muscles, one predomitly in skeletal (a-skeletal) and one in cardiac (a-cardiac) muscle cells. These actin isoforms differ slightly in their N-terminus and the sequences of each of them are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown to appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases.
Especificaciones
Antígeno Alpha SMA
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.35 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica ACTA2
N.º de referencia del gen P62736, P62737, P62738
Alias de gen a SMA, AAT6, ACTA2, ACTA2 Smooth muscle Actin, ACTA2 Smooth muscle Actin Specific, Actin, aortic smooth muscle, Alpha actin 2, Alpha SMA, Alpha SMA Specific, a-SMA, smooth muscle actin, alpha SMA
Símbolos de los genes ACTA2
Especie del huésped Rabbit
Inmunógeno Peptide
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11475, 59, 81633
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG