Learn More
alpha 2u-Globulin Mouse anti-Rat, Clone: 129736, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 1 publication
Marca:  R&D Systems MAB586-SP
Encompass
Código de producto. 15707674
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »
Descripción
Descripción
alpha 2u-Globulin Monoclonal specifically detects alpha 2u-Globulin in Rat samples. It is validated for Immunohistochemistry.
Especificaciones
Especificaciones
alpha 2u-Globulin
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
259246
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Immunohistochemistry
129736
Immunohistochemistry 8-25 ug/mL
AUG, Mup, PGCL1
Rat urine-derived alpha 2u -Globulin
25 μg
Primary
Detects rat alpha 2μ -Globulin in direct ELISAs.
Rat
Purified
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones