missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALOX12 Monoclonal Antibody (OTI5H2), TrueMAB™, OriGene
Description
ALOX12 Monoclonal antibody specifically detects ALOX12 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | ALOX12 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI5H2 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | PRKG2 |
| N.º de referencia del gen | Q13237 |
| Alias de gen | 12(S)-lipoxygenase; 12LO; 12-LO; 12LOX; 12-LOX; 12S LOX; 12S-lipoxygenase; 12S-LOX; 9930022G08Rik; ALOX12; Alox12p; arachidonate 12-lipoxygenase; arachidonate 12-lipoxygenase, 12S type; arachidonate 12-lipoxygenase, 12S-type; Arachidonate 15-lipoxygenase,15S-type; Linoleate 13S-lipoxygenase; Lipoxin synthase 12-LO; LOG12; P-12LO; platelet 12-LOX; platelet-type 12-lipoxygenase; platelet-type lipoxygenase 12 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?