Learn More
Albendazole, TRC
Código de producto. 18050030
Change view
Click to view available options
Cantidad:
100 mg
1 g
10 g
Tamaño de la unidad:
1 g
10 g
100 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
18050030 100 mg 100 miligramos
30138055 10 g 10 g
30136939 1 g 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 18050030 Proveedor Toronto Research Chemicals N.º de proveedor TRCA511610100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
CAS 54965-21-8
Nombre del producto químico o material Albendazole
Formula Weight (peso de la fórmula) 265.0885
Fórmula InChI InChI=1S/C12H15N3O2S/c1-3-6-18-8-4-5-9-10(7-8)14-11(13-9)15-12(16)17-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)
Nombre IUPAC methyl N-(5-propylsulfanyl-1H-benzimidazol-2-yl)carbamate
Fórmula molecular C12 H15 N3 O2 S
Peso molecular (g/mol) 265.33
Porcentaje de pureza >95%
Notas de grado de pureza HPLC
SMILES CCCSc1ccc2[nH]c(NC(=O)OC)nc2c1
Sinónimo Carbamic acid, [5-(propylthio)-1H-benzimidazol-2-yl]-, methyl ester (9CI), Albamelin, Alben, Albendazole, Albendol, Albenza, Albenzil, Aldifal, Alminth, Alvet, Alzental, Andazol, Ashialben, Atasol, Baxen, Bruzol, Eskazole, Exiptol, Haojing, Helminsols, Loveral, Lurdex, Methyl 5-propylthio-2-benzimidazolecarbamate, NSC 220008, SKF 62979, Tobend, Valbazen, Vermisen, Vermitan, Vermizole, Vetalben, Zentel