missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKAP17A Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA521413
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: 293T, HeLaS3, Raji. Predicted reactivity: Rhesus Monkey (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DXYS155E is a gene found in the pseudoautosomal region of the distal short arms of the X and Y chromosomes, and appears to be ubiquitously expressed.
Especificaciones
| AKAP17A | |
| Polyclonal | |
| Unconjugated | |
| AKAP17A | |
| 721P; A kinase (PRKA) anchor protein 17A; AKAP17A; AKAP-17A; A-kinase anchor protein 17A; A-kinase anchoring protein 17A; B-lymphocyte antigen; B-lymphocyte surface antigen; CCDC133; CXYorf3; DXYS155E; PRKA17A; Protein kinase A-anchoring protein 17A; Protein XE7; pseudoautosomal gene XE7; RP13-297E16.1; SFRS17A; Splicing factor, arginine/serine-rich 17A; XE7; XE7Y | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8227 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q02040 | |
| AKAP17A | |
| Recombinant protein encompassing a sequence within the C-terminus region of human SFRS17A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido