Learn More
Notrax Airug™ Construction of Microcellular Vinyl with Ribbed Pattern, Black
Notrax Airug™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles.
Marca:  Notrax 410C0348BL
Detalles adicionales : Peso : 2.80000kg
 Ver más versiones de este producto
Código de producto. 17792058
  • 62.12€ / cada uno
Fecha estimada de envío: 03-04-2023
para ver el stock
Añadir a la cesta
Descripción
Descripción
  • Ribbed pattern enhances traction
  • Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
  • All four sides are bevelled and sealed to minimize tripping hazards
Especificaciones
Especificaciones
122 cm
Microcellular Vinyl
60 ft.
Industrial and Commercial Matting
1830 cm
Black