Learn More
Invitrogen™ AID Monoclonal Antibody (1AID-2E11)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA3081
Descripción
MA3-081 detects AID from human, and mouse samples. MA3-081 has been successfully used in Western blot, immunofluorescence, flow cytometry, and ELISA applications. The MA3-081 immunogen is the synthetic peptide coupled to ovalalbumin, aa 185 to 198. MA3-081 detects Isoform 1 (~24kDa) and Isoform 2 (~22kDa) in human samples.
This gene encodes a RNA-editing deaminase that is a member of the cytidine deaminase family. The protein is involved in somatic hypermutation, gene conversion, and class-switch recombination of immunoglobulin genes. Defects in this gene are the cause of autosomal recessive hyper-IgM immunodeficiency syndrome type 2 (HIGM2).
Especificaciones
| AID | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.05% sodium azide | |
| Q9GZX7, Q9WVE0 | |
| Aicda | |
| Synthetic peptide coupled to Ovalbumin corresponding to aa 185 to 198 of mouse AID. | |
| RUO | |
| 11628, 57379 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 1AID-2E11 | |
| Unconjugated | |
| Aicda | |
| activation induced deaminase; activation-induced cytidine deaminase; Aicda; AID; Arp2; CDA2; Cytidine aminohydrolase; epididymis secretory protein Li 284; HEL-S-284; HIGM2; integrated into Burkitt's lymphoma cell line Ramos; single-stranded DNA cytosine deaminase | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.